Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sulphuric acid
Sulphuric acid 0.1 mol concentrated aqueous solution volumetric solution
Sulphuric acid 0.1 mol concentrated aqueous solution volumetric solution
CHMP165750641.0001
:
CHEMPUR
:
Sulphuric acid 0.1 mol concentrated aqueous solution volumetric solution
CHMP165750641.0001
:
CHEMPUR
:
Pk:
1
Seq:
0001
:
:
98.08 g/mol
:
Ambient
:
231-639-5
:
2796
:
8,II
:
GHS05
:
7664-93-9
:
H₂SO₄
:
MFCD00064589