
Copper(II) sulphate 0.05 mol/l volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7758-98-7
:
Formula:CuSO₄
MDL Number:MFCD00010981
:159.61 g/mol
EINECS:231-847-6
:GHS09
Storage Temperature:Ambient
Frequently Bought Together