Order Entry
Poland
ContactUsLinkComponent
 
Hydrogen peroxide 45%, pure analytical reagent
Catalog # CHMP118851930.1000
Supplier:  CHEMPUR
CAS Number:  
Hydrogen peroxide 45%, pure analytical reagent
Catalog # CHMP118851930.1000
Supplier:  CHEMPUR
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

About this item

 :
Boiling Pt:126 °C (1013 hPa)
Melting Pt:–40 °C
Density:1.19 g/cm³ (20 °C)
ADR:5.1,II
 :34.01 g/mol
EINECS:231-765-0
UN:2014
 :GHS03
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333