
Potassium permanganate 0.1 mol concentrated aqueous solution volumetric solution
Catalog # CHMP167438909.0001
Supplier: CHEMPUR
CAS Number:
Specifications
- Pk:1
Specifications
About this item
:
:158.03 g/mol
Storage Temperature:Ambient
EINECS:231-760-3
UN:3082
ADR:9,III
:GHS09
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364