Vanillin, pure analytical reagent
Glass bottle for solids, 100 g
POCH874180115-100G
: 4-Hydroxy-3-methoxybenzaldehyde
- :121-33-5
- :C₈H₈O₃
- :MFCD00006942
- :152.15 g/mol
- :13,09998
- :285 °C (1013 hPa)
- :∼82 °C
- :1.056 g/cm³ (20 °C)
- :153 °C
- :Ambient
- :204-465-2
- Pack type:Glass bottle for solids
- Pk:100 g