Magnesium chloride hexahydrate, pure analytical reagent
Supplier: POCH AVANTOR BRAND
Certificates
About this item
CAS Number:7791-18-6
:
Formula:MgCl₂·6H₂O
MDL Number:MFCD00149781
:203.3 g/mol
Merck Index:13,05684
Boiling Pt:1412 °C (1013 hPa)
Melting Pt:117 °C
Density:1.57 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:232-094-6
Frequently Bought Together