
Sulphuric acid 2 mol/l (4 N) in aqueous solution volumetric solution
: CHEMPUR
:7664-93-9
:
:H₂SO₄
:MFCD00064589
:98.08 g/mol
:1.146 g/cm³ (20 °C)
:Ambient
:231-639-5
:2796
:8,II
:GHS05
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
Accessories for VWR® Pipette Controllers
: VWR Collection
VWR®, Graduated Pipettes, Type 3, Class B
: VWR Collection
Glycerine, anhydrous, pure analytical reagent
: POCH AVANTOR BRAND
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Sodium chloride, tablets
: CHEMPUR