Sulphuric acid 0.1 mol/l (0.2 N) in aqueous solution
Supplier: POCH AVANTOR BRAND
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
Frequently Bought Together