Hydrochloric acid 0.2 mol/l (0.2 N) volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1.004 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,III
:36.46 g/mol
Frequently Bought Together