Hydrogen peroxide 60%, pure
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7722-84-1
:
Formula:H₂O₂
MDL Number:MFCD00011333
Boiling Pt:126 °C (1013 hPa)
Melting Pt:–40 °C
Density:1.19 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-765-0
UN:2014
ADR:5.1,II
:34.01 g/mol
:GHS03
:GHS05
:GHS07
Frequently Bought Together