

Hydrogen peroxide 30%, pure analytical reagent
: CHEMPUR
:7722-84-1
:
:H₂O₂
:MFCD00011333
:∼107 °C (1013 hPa)
:< 0 °C
:1.11 g/cm³ (20 °C)
:231-765-0
:2014
:5.1,II
:34.01 g/mol
:GHS05
:GHS07
:Ambient
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methyl red indicator
: CHEMPUR
Ethanol, GPR RECTAPUR®
: VWR Chemicals