Hydrogen peroxide 3%, pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7722-84-1
:
Formula:H₂O₂
MDL Number:MFCD00011333
Boiling Pt:100 °C (1013 hPa)
Density:1.01 g/cm³ (20 °C)
:34.01 g/mol
Storage Temperature:Ambient
EINECS:231-765-0
Frequently Bought Together