Hydrogen peroxide 10%, pure analytical reagent
Supplier: CHEMPUR
Jump to:
About this item
- CAS Number:7722-84-1
- Formula:H₂O₂
- MDL Number:MFCD00011333
- Molecular Weight:34.01 g/mol
- Boiling Pt:150.2 °C (1013 hPa)
- Melting Pt:–33 °C
- Density:1.11 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-765-0
- UN:2984
- ADR:5.1,III
- :Density: 1.11 g/cm³ (20 °C)
2 Options Available Below
Product Details & Documents
- CAS Number:7722-84-1
- Formula:H₂O₂
- MDL Number:MFCD00011333
- Molecular Weight:34.01 g/mol
- Boiling Pt:150.2 °C (1013 hPa)
- Melting Pt:–33 °C
- Density:1.11 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-765-0
- UN:2984
- ADR:5.1,III
- :Density: 1.11 g/cm³ (20 °C)
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



