

Perchloric acid 65%, pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7601-90-3
:
Formula:HClO₄
MDL Number:MFCD00011325
:100.46 g/mol
Boiling Pt:160…181 °C (1013 hPa)
Melting Pt:–112.2 °C
Density:1.53…1.768 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-512-4
UN:1873
ADR:5.1,I
:GHS03
:GHS05
Frequently Bought Together