

Copper(II) sulphate 0.5 mol/l volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7758-98-7
:
Formula:CuSO₄
MDL Number:MFCD00010981
:159.61 g/mol
Storage Temperature:Ambient
EINECS:231-847-6
UN:3082
ADR:9,III
:GHS07
:GHS09
Frequently Bought Together