Celite® 545, filter aid
Glass bottle for solids, 250 g
Catalog # POCH213500460-250G
Supplier: POCH AVANTOR BRAND
Celite® 545, filter aid
Catalog # POCH213500460-250G
Supplier: POCH AVANTOR BRAND
CAS Number: 68855-54-9
Jump to:
Product Details & Documents
Synonyms: DE, Diatomaceous earth, flux-calcined
- CAS Number:68855-54-9
- Formula:SiO₂
- MDL Number:MFCD00132803
- Molecular Weight:60.08 g/mol
- EINECS:272-489-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



