Order Entry
Poland
Orders LinkContactUsLinkComponent
Copper(II) carbonate basic, pure

Copper(II) carbonate basic, pure

Irritant
Supplier:   CHEMPUR
Copper(II) carbonate basic, pure
Supplier:  CHEMPUR
Certificates
Copper(II) carbonate basic, pure

3 Options Available Below

Product Details & Documents
Synonyms:  Copper (II) carbonate dihydroxide, Copper(II) carbonate, basic, Basic copper carbonate, Copper (II) carbonate-copper (II) hydroxide (1:1)
  • CAS Number:12069-69-1
  • Formula:CuCO₃·Cu(OH)₂
  • MDL Number:MFCD00010976
  • Molecular Weight:221.12 g/mol
  • Merck Index:13,02658
  • Melting Pt:200 °C
  • Density:4 g/cm³ (20 °C)
  • EINECS:235-113-6
  • UN:3288
  • ADR:6.1,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.