0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
Sulphuric acid
Sulphuric acid 0.25 mol concentrated aqueous solution volumetric solution
Sulphuric acid 0.25 mol concentrated aqueous solution volumetric solution
CHMP165750643.0001
:
CHEMPUR
:
Sulphuric acid 0.25 mol concentrated aqueous solution volumetric solution
CHMP165750643.0001
:
CHEMPUR
:
Pk:
1
Seq:
0001
:
:
98.08 g/mol
:
Ambient
:
231-639-5
:
2796
:
8,II
:
GHS05
:
7664-93-9
:
H₂SO₄
:
MFCD00064589