Order Entry
Poland
ContactUsLinkComponent
N,N-Dimethylformamide for DNA and peptides synthesis
N,N-Dimethylformamide for DNA and peptides synthesis
Catalog # POCH355123699-1L
Supplier:  POCH AVANTOR BRAND
CAS Number:  
undefined
N,N-Dimethylformamide for DNA and peptides synthesis
Catalog # POCH355123699-1L
Supplier:  POCH AVANTOR BRAND
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
 :
 :73.09 g/mol
EINECS:200-679-5
UN:2265
REACH:01-2119475605-32
 :GHS02
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284
Boiling Pt:153
Melting Pt:–61 °C
Density:0.949 g/cm³ (20 °C)
Flash Pt:57.5 °C (closed cup)
ADR:3 III
Merck Index:13,03269