



Glutaraldehyde 25% in aqueous solution, pure
Supplier: CHEMPUR
Certificates
About this item
CAS Number:111-30-8
:
Formula:C₅H₈O₂
MDL Number:MFCD00007025
:100.12 g/mol
Boiling Pt:101 °C (1013 hPa)
Melting Pt:–6 °C
Density:1.06 g/cm³ (20 °C)
Flash Pt:>71 °C
EINECS:203-856-5
UN:2927
ADR:6.1,II
:GHS06
:GHS05
:GHS08
:GHS09
Storage Temperature:Refrigerator
Frequently Bought Together