Copper(II) sulphate, anhydrous, pure analytical reagent
Supplier: POCH AVANTOR BRAND
Certificates
About this item
Synonyms: Cupric sulphate
CAS Number:7758-98-7
:
Formula:CuSO₄
MDL Number:MFCD00010981
Density:3.603 g/cm³ (25 °C)
Molecular Weight:159.61 g/mol
Melting Pt:200
ADR:6.1 III
Merck Index:12,02722
EINECS:231-847-6
UN:3288
:GHS07
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together