
Hydrochloric acid 0.2 mol concentrated aqueous solution volumetric solution
Catalog # CHMP165753137.0001
Supplier: CHEMPUR
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,II
:36.46 g/mol
UN:1789
:GHS05
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324