Order Entry
Poland
Orders LinkContactUsLinkComponent
Cream butyrometer, without stopper
Cream butyrometer, without stopper
Supplier:   GERBER FUNKE
Cream butyrometer, without stopper
Supplier:  GERBER FUNKE
Certificates
Cream butyrometer, without stopper
Cream butyrometer, without stopper
4 Options Available Below
Product Details & Documents
According to Roeder´s weighing method.
Product Family Options
Product Information
  • Availability
    Price
  • 614-0108
    Butyrometer for cream, Roeder´s, 0 - 30 - 55 % : 0,5
    Butyrometer for cream, Roeder´s, 0 - 30 - 55 % : 0,5
    Log in for availability
    Log in for availability
    1 * 1 SZT
    1 * 1 SZT - 131 zł
    131 zł
    1 * 1 SZT
       131 zł
  • 614-0118
    Butyrometer for cream, Roeder´s, 0 - 5 - 70 % : 1,0
    Butyrometer for cream, Roeder´s, 0 - 5 - 70 % : 1,0
    Log in for availability
    Log in for availability
    1 * 10 SZT
    1 * 10 SZT - 719 zł
    719 zł
    1 * 10 SZT
       719 zł
  • 614-0117
    Butyrometer for cream, Roeder´s, 0 - 50 - 75 % : 0,5
    Butyrometer for cream, Roeder´s, 0 - 50 - 75 % : 0,5
    Log in for availability
    Log in for availability
    1 * 10 SZT
    1 * 10 SZT - 999 zł
    999 zł
    1 * 10 SZT
       999 zł
  • 614-0107
    Butyrometer for cream, Roeder´s, 0 - 5 - 40 % : 0,5
    Butyrometer for cream, Roeder´s, 0 - 5 - 40 % : 0,5
    Log in for availability
    Log in for availability
    1 * 10 SZT
    1 * 10 SZT - 607 zł
    607 zł
    1 * 10 SZT
       607 zł
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Weighing boat for butter for butyrometers acc. to Roeder's method
    Weighing boat for butter for butyrometers acc. to Roeder's method
    614-2034
    Log in for availability
    1 * 1 SZT - 22,10 zł
Product Information
  • Catalog #
    Availability
    Price
  • Cream beaker, unperforated, for ice cream and condensed milk butyrometer and cream butyrometer acc. to Roeder´s method
    Cream beaker, unperforated, for ice cream and condensed milk butyrometer and cream butyrometer acc. to Roeder´s method
    614-2032
    Log in for availability
    1 * 1 SZT - 8,30 zł
Product Information
  • Catalog #
    Availability
    Price
  • Stopper, rubber, with hole, for all weighing method butyrometers
    Stopper, rubber, with hole, for all weighing method butyrometers
    614-2030
    Log in for availability
    1 * 100 SZT - 306 zł
Product Information
  • Catalog #
    Availability
    Price
  • Stopper, rubber, for sealing the bulbs of all weighing method butyrometers
    Stopper, rubber, for sealing the bulbs of all weighing method butyrometers
    614-2029
    Log in for availability
    1 * 100 SZT - 102 zł