Order Entry
Poland
ContactUsLinkComponent
Sodium standard solution, concentrate, 1000 mg Na in water (from NaCl) Titrisol® standard for AAS, Supelco®
Sodium standard solution, concentrate, 1000 mg Na in water (from NaCl) Titrisol® standard for AAS, Supelco®
Catalog # 1.09927.0001
Dostawca:  Merck
Numer CAS:  
Sodium standard solution, concentrate, 1000 mg Na in water (from NaCl) Titrisol® standard for AAS, Supelco®
Catalog # 1.09927.0001
Dostawca:  Merck
Numer CAS:  

Dane techniczne

  • Concentration:
    1000 mg
  • Matrix:
    H₂O
  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • M-Clarity quality level:
    MQ100
  • Accuracy:
    ±2 mg
  • Element:
    Sodium
  • Symbol:
    Na

Dane techniczne

Informacje o tym produkcie

Certifications: Traceable to SRM from NIST