Order Entry
Poland
ContactUsLinkComponent
 
Gold standard solution, concentrate, 1000 mg Au in 12,7% hydrochloric acid (from HAuCl?) Titrisol® standard for AAS, Supelco®
Catalog # 1.09868.0001
Supplier:  Merck
CAS Number:  
Gold standard solution, concentrate, 1000 mg Au in 12,7% hydrochloric acid (from HAuCl?) Titrisol® standard for AAS, Supelco®
Catalog # 1.09868.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • Element:
    Gold
  • Symbol:
    Au

Specifications

About this item

Certifications: Traceable to SRM from NIST