
(R)-(+)-1,1'-Bi-2-naphthol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18531-94-7
:
Formula:C₂₀H₁₄O₂
MDL Number:MFCD00004068
:286.33 g/mol
Melting Pt:205…211 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to light beige Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:205 to 211 °C
- HPLC:≥98.5 %
- Specific optical rotation:+33° to +38° (20 °C, 589 nm) (c=1, THF)
Specification Test Results
Frequently Bought Together