




Ethylaluminium dichloride 0.9 M in heptane, AcroSeal®
:563-43-9
:
:C₂H₅AlCl₂
:MFCD00000457
:126.95 g/mol
:0.74 g/cm³
:Ambient
:209-248-6
:3399
:4.3,I
:GHS07
:GHS08
:GHS05
:GHS09
:GHS02
- Appearance:Clear colorless Solution
- Assay:14.5 to 15.5 %
- Loss on drying:≤4 %
- HPLC:≥97.5 % (mixture of A and B)
Frequently Bought Together