Order Entry
Poland
ContactUsLinkComponent
 
Sarcosine methyl ester hydrochloride, Sigma-Aldrich®
Catalog # SIAL84570-50G
Supplier:  Merck
CAS Number:  
Sarcosine methyl ester hydrochloride, Sigma-Aldrich®
Catalog # SIAL84570-50G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :139.58 g/mol
EINECS:236-853-2
CAS Number:13515-93-0
Formula:C₄H₉NO₂·HCl
MDL Number:MFCD00038876