Order Entry
Poland
ContactUsLinkComponent
Zirconium standard solution, 1000 mg/L Zr in 5% hydrofluoric acid standard for AAS
Zirconium standard solution, 1000 mg/L Zr in 5% hydrofluoric acid standard for AAS
  ACRO196471000
 :  
Zirconium standard solution, 1000 mg/L Zr in 5% hydrofluoric acid standard for AAS
  ACRO196471000
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    5% HF
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Zirconium
  • Symbol:
    Zr