Order Entry
Poland
ContactUsLinkComponent
Sodium standard solution, 1000 mg/L Na in water (from NaNO₃) Certipur® standard for AAS, Supelco®
Sodium standard solution, 1000 mg/L Na in water (from NaNO₃) Certipur® standard for AAS, Supelco®
Catalog # 1.19507.0500
Supplier:  Merck
CAS Number:  
Sodium standard solution, 1000 mg/L Na in water (from NaNO₃) Certipur® standard for AAS, Supelco®
Catalog # 1.19507.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • M-Clarity quality level:
    MQ100
  • Accuracy:
    ±10 mg/l
  • Element:
    Sodium
  • Symbol:
    Na

Specifications

About this item

Certifications: Traceable to SRM from NIST