Order Entry
Poland
ContactUsLinkComponent
TISAB III solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations
TISAB III solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations
Catalog # METT51350106
Supplier:  Mettler - Toledo
CAS Number:  
undefined
TISAB III solution (Total Ionic Strength Adjustment Buffer) for fluoride determinations
Catalog # METT51350106
Supplier:  Mettler - Toledo
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Description:
    TISAB III solution for fluoride determinations
  • pH:
    4,5

Specifications

About this item