Potassium chloride 0.1 mol concentrated aqueous solution volumetric solution
Catalog # CHMP161397402.0001
Supplier: CHEMPUR
CAS Number:
Specifications
- Pk:1
Specifications
About this item
:
:74.55 g/mol
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360