Ethylene glycol 10% in aqueous solution, Technical Grade
Supplier: CHEMPUR
Certificates
About this item
CAS Number:107-21-1
:
Formula:HOCH₂CH₂OH
MDL Number:MFCD00002885
:62.07 g/mol
Storage Temperature:Refrigerator
EINECS:203-473-3
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Frequently Bought Together