Sodium acetate 0.05 mol/l (0.05 N) in aqueous solution volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:127-09-3
:
Formula:H₃CCOONa
MDL Number:MFCD00012459
:82.03 g/mol
Storage Temperature:Ambient
EINECS:204-823-8
Frequently Bought Together