Simazine(2-chloro-4,6-bis(ethylamino)-1,3,5-triazine) ≥99.5%
Catalog # 124502D
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass ampoule
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:201.66 g/mol
Melting Pt:219 °C
Flash Pt:100 °C (closed cup)
EINECS:204-535-2
UN:3077
ADR:9,III
:GHS08
:GHS09
CAS Number:122-34-9
Formula:C₇H₁₂ClN₅
MDL Number:MFCD00023174