Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Barium chloride
Barium chloride 0.1 mol concentrated aqueous solution volumetric solution
Barium chloride 0.1 mol concentrated aqueous solution volumetric solution
Barium chloride 0.1 mol concentrated aqueous solution volumetric solution
CHMP161579709.0001
:
CHEMPUR
:
Barium chloride 0.1 mol concentrated aqueous solution volumetric solution
CHMP161579709.0001
:
CHEMPUR
:
Pk:
1
:
:
208.24 g/mol
:
1.18 g/cm³ (20 °C)
:
Ambient
:
233-788-1
:
3287
:
6.1,III
:
GHS07
:
10361-37-2
:
BaCl₂
:
MFCD00003445