Order Entry
Poland
ContactUsLinkComponent
Phthaloyl dichloride for synthesis, Sigma-Aldrich®
Phthaloyl dichloride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Phthaloyl dichloride for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    16 °C
  • Molar Mass:
    203.03 g/mol
  • Boiling Point:
    281 °C
  • Flash Point:
    110 °C
  • Refractive Index:
    1.5684 (20 °C, 589 nm)
  • Density:
    1.41 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together