Sodium thiosulphate, anhydrous, pure analytical reagent
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7772-98-7
:
Formula:Na₂S₂O₃
MDL Number:MFCD00003499
:158.11 g/mol
Merck Index:12,08844
Boiling Pt:100 °C (1013 hPa)
Melting Pt:48 °C
Density:1.66 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-867-5
Frequently Bought Together