Erbium triacetate tetrahydrate ≥99% (REO, rare earth oxide basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15280-57-6
:
Formula:C₆H₉ErO₆·4H₂O
MDL Number:MFCD00013034
:416.45 g/mol
Density:2.114
Storage Temperature:Ambient
EINECS:247-067-4
Related Information
Frequently Bought Together