

Tetrachloroethylene, pure analytical reagent
Catalog # POCH275920114-1L
Supplier: POCH AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:165.83 g/mol
Merck Index:12,09332
Boiling Pt:121.1 °C (1013 hPa)
Melting Pt:–22 °C
Density:1.63 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-825-9
UN:1897
ADR:6.1,III
:GHS08
:GHS09
CAS Number:127-18-4
Formula:C₂Cl₄
MDL Number:MFCD00000834