2-Propanol for spectroscopy
POCH751500830-2.5L
:
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
:
:60.1 g/mol
:12
:200-661-7
:1219
:01-2119457558-25
:GHS02
:GHS07
:Ambient
:67-63-0
:(CH₃)₂CHOH
:MFCD00011674
:82
:–89 °C
:0.786 g/cm³ (20 °C)
:3 II
:13,05228