


1-Propanol, pure analytical reagent
Catalog # POCH751460114-1L
Supplier: POCH AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:60.1 g/mol
Merck Index:13,07934
Boiling Pt:97 °C (1013 hPa)
Melting Pt:–127 °C
Density:0.8053 g/cm³ (20 °C)
Flash Pt:15 °C
EINECS:200-746-9
UN:1274
ADR:3,II
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:71-23-8
Formula:H₃CCH₂CH₂OH
MDL Number:MFCD00002941