Isoamyl acetate, pure
: CHEMPUR
:123-92-2
:
:C₇H₁₄O₂
:MFCD00008946
:130.19 g/mol
:12,05125
:142 °C (1013 hPa)
:–78 °C
:0.87 g/cm³ (20 °C)
:25 °C
:Ambient
:204-662-3
:1104
:3,III
:GHS02
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Frequently Bought Together