

Acetone for gas chromatography, for pesticide residue analysis
CHMP611024802.1000
: CHEMPUR
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
:
:58.08 g/mol
:13,00067
:56.2 °C (1013 hPa)
:–95.4 °C
:0.792 g/cm³ (20 °C)
:<–20 °C (closed cup)
:Ambient
:200-662-2
:1090
:3,II
:01-2119471330-49
:GHS02
:GHS07
:67-64-1
:CH₃COCH₃
:MFCD00008765