Calcium chloride 0.1 mol/l (0.2 N) in aqueous solution volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:10043-52-4
:
Formula:CaCl₂
MDL Number:MFCD00010903
:110.98 g/mol
Storage Temperature:Ambient
EINECS:233-140-8
Frequently Bought Together