Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7772-98-7
:
Formula:Na₂S₂O₃
MDL Number:MFCD00003499
:158.11 g/mol
Storage Temperature:Ambient
EINECS:231-867-5
Frequently Bought Together
Sulphuric acid 95,0 - 97,0% 95.0-97.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR®, Single-Channel Pipettes, Mechanical, Variable Volume
Supplier: VWR Collection
Hydrochloric acid 0.1 mol/l (0.1 N), AVS TITRINORM Reag. Ph. Eur. volumetric solution
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Chemical spoon with spatula end
Supplier: BOCHEM
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ammonium iron(III) sulphate dodecahydrate, pure analytical reagent
Supplier: CHEMPUR
Formic acid ≥80%, pure analytical reagent
Supplier: CHEMPUR
Potassium iodide, pure analytical reagent
Supplier: CHEMPUR
Phenolphthalein 1% in ethanol
Supplier: CHEMPUR