
Sodium hydroxide 1 mol/l (1 N) in aqueous solution volumetric solution
: CHEMPUR
:1310-73-2
:
:NaOH
:MFCD00003548
:40 g/mol
:1.04 g/cm³ (25 °C)
:Ambient
:215-185-5
:3266
:8,III
:GHS05
Frequently Bought Together
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
: VWR Chemicals
n-Pentane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Benzyl alcohol ≥99.0% analytical reagent
: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals