
Nitric acid 0.1 mol/l (0.1 N) volumetric solution
Supplier: CHEMPUR
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
UN:3264
ADR:8,III
:GHS05
Frequently Bought Together