Nitric acid concentrate for dilution to 0.1 mol in aqueous solution volumetric solution
Catalog # CHMP165296834.0001
Supplier: CHEMPUR
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:63.01 g/mol
Density:1.38 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
:GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349