Order Entry
Poland
Orders LinkContactUsLinkComponent
Sodium hydroxide 45% in aqueous solution 45% in aqueous solution, pure
 :   CHEMPUR
Sodium hydroxide 45% in aqueous solution 45% in aqueous solution, pure
 :  CHEMPUR
Sodium hydroxide 45% in aqueous solution 45% in aqueous solution, pure
 :  Caustic soda
  •  :1310-73-2
  •  :NaOH
  •  :MFCD00003548
  •  :40 g/mol
  •  :119…135 °C (1013 hPa)
  •  :9…10 °C
  •  :1.53 g/cm³ (20 °C)
  •  :Ambient
  •  :215-185-5
  •  :1824
  •  :8,II
Product Family Options